Assay Details
| Gene Symbol: RB1CC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011517643.2 | XP_011515945.1 | 3 - 4 | 702 | 143 | |
| XM_017014104.1 | XP_016869593.1 | 3 - 4 | 733 | 143 | ||
| XM_017014107.1 | XP_016869596.1 | 3 - 4 | 707 | 143 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011517643.2, XM_017014104.1, XM_017014107.1 transcripts.
Set Membership: |
|

RB1CC1
Genomic Map