Assay Details
| Gene Symbol: TGFBRAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142621.2 | NP_001136093.1 | 7 - 8 | 1627 | 88 | |
| NM_004257.5 | NP_004248.2 | 7 - 8 | 1630 | 88 | ||
| XM_011512193.2 | XP_011510495.1 | 7 - 8 | 1587 | 88 | ||
| XM_017005339.1 | XP_016860828.1 | 6 - 7 | 1123 | 88 | ||
| GenBank mRNA | AF022795.1 | - | 6 - 7 | 1565 | 88 | |
| AK021697.1 | - | 5 - 6 | 688 | 88 | ||
| AK291382.1 | - | 7 - 8 | 1595 | 88 | ||
| BC020548.2 | - | 7 - 8 | 1588 | 4420120 | 88 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK021697.1 transcripts.
Set Membership: |
|

TGFBRAP1
Genomic Map