Assay Details
| Gene Symbol: PUM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005262607.1 | XP_005262664.1 | 16 - 17 | 2738 | 94 | |
| XM_005262609.1 | XP_005262666.1 | 15 - 16 | 2501 | 94 | ||
| XM_006711973.1 | XP_006712036.1 | 16 - 17 | 2535 | 94 | ||
| XM_006711974.2 | XP_006712037.1 | 16 - 17 | 2575 | 94 | ||
| XM_006711975.2 | XP_006712038.1 | 17 - 18 | 2699 | 94 | ||
| XM_011532719.1 | XP_011531021.1 | 18 - 19 | 2934 | 94 | ||
| XM_011532720.2 | XP_011531022.1 | 18 - 19 | 2943 | 94 | ||
| XM_011532721.2 | XP_011531023.1 | 17 - 18 | 2735 | 94 | ||
| XM_011532723.1 | XP_011531025.1 | 15 - 16 | 2365 | 94 | ||
| XM_017003703.1 | XP_016859192.1 | 17 - 18 | 2763 | 94 | ||
| XM_017003704.1 | XP_016859193.1 | 19 - 20 | 3081 | 94 | ||
| XM_017003705.1 | XP_016859194.1 | 19 - 20 | 3068 | 94 | ||
| XM_017003706.1 | XP_016859195.1 | 18 - 19 | 2860 | 94 | ||
| XM_017003707.1 | XP_016859196.1 | 15 - 16 | 2298 | 94 | ||
| XM_017003708.1 | XP_016859197.1 | 15 - 16 | 2531 | 94 | ||
| GenBank mRNA | BC024218.2 | - | 4 - 5 | 761 | 94 | |
More Information
Set Membership: |
|

PUM2
Genomic Map