Assay Details
| Gene Symbol: STARD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014725.4 | NP_055540.2 | 3 - 4 | 358 | 72 | |
| GenBank mRNA | BC035587.1 | - | 3 - 4 | 369 | 5518429 | 72 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC035587.1, NM_014725.4 transcripts.
Set Membership: |
|

STARD8
Genomic Map