Assay Details
| Gene Symbol: NAV2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244963.1 | NP_001231892.1 | 6 - 7 | 1199 | 82 | |
| XM_006718365.3 | XP_006718428.1 | 6 - 7 | 1521 | 82 | ||
| XM_006718366.3 | XP_006718429.1 | 6 - 7 | 685 | 82 | ||
| XM_011520444.2 | XP_011518746.1 | 6 - 7 | 1521 | 82 | ||
| XM_011520445.2 | XP_011518747.1 | 6 - 7 | 1521 | 82 | ||
| XM_011520446.2 | XP_011518748.1 | 6 - 7 | 1521 | 82 | ||
| XM_011520447.2 | XP_011518749.1 | 6 - 7 | 1521 | 82 | ||
| XM_011520448.2 | XP_011518750.1 | 6 - 7 | 1392 | 82 | ||
| XM_011520449.2 | XP_011518751.1 | 6 - 7 | 1521 | 82 | ||
| XM_011520450.2 | XP_011518752.1 | 6 - 7 | 1521 | 82 | ||
| XM_017018520.1 | XP_016874009.1 | 7 - 8 | 1313 | 82 | ||
| XM_017018521.1 | XP_016874010.1 | 6 - 7 | 1521 | 82 | ||
| XM_017018523.1 | XP_016874012.1 | 6 - 7 | 2198 | 82 | ||
| XM_017018524.1 | XP_016874013.1 | 6 - 7 | 1021 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000252.hg.1
Set Membership: |
|

NAV2
Genomic Map