Assay Details
| Gene Symbol: SYNM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015286.5 | NP_056101.5 | 4 - 5 | 3570 | 79 | |
| GenBank mRNA | AF359284.1 | - | 4 - 5 | 3561 | 79 | |
| AJ310521.1 | - | 4 - 5 | 3564 | 79 | ||
| AL833262.1 | - | 4 - 5 | 3061 | 79 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000953.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008845.hg.1 | ENST00000328642.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008845.hg.1 | ENST00000336292.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008845.hg.1 | ENST00000560674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008845.hg.1 | ENST00000561323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008845.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008845.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008845.hg.1 | TC15000953.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010610.hg.1 | ENST00000566974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010610.hg.1 | ENST00000566974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183730.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183730.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183730.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183730.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183730.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183730.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183730.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183730.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183730.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183730.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183730.hg.1 | TC1500010796.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500010610.hg.1 | ENST00000566974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010610.hg.1 | ENST00000566974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183729.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183729.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183729.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183729.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183729.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183729.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183729.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183729.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183729.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183729.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183729.hg.1 | TC1500010796.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_145728.2 | Refseq |

SYNM
Genomic Map