Assay Details
| Gene Symbol: TMEM245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032012.3 | NP_114401.2 | 9 - 10 | 1567 | 129 | |
| XM_011518446.2 | XP_011516748.1 | 9 - 10 | 1582 | 129 | ||
| XM_011518447.1 | XP_011516749.1 | 8 - 9 | 1543 | 129 | ||
| XM_011518449.1 | XP_011516751.1 | 8 - 9 | 1450 | 129 | ||
| XM_011518452.1 | XP_011516754.1 | 8 - 9 | 1333 | 129 | ||
| XM_017014571.1 | XP_016870060.1 | 8 - 9 | 1540 | 129 | ||
| XM_017014572.1 | XP_016870061.1 | 7 - 8 | 1309 | 129 | ||
| GenBank mRNA | AF153415.1 | - | 9 - 10 | 1567 | 129 | |
| AK172815.1 | - | 3 - 4 | 312 | 129 | ||
| AK299952.1 | - | 9 - 10 | 1549 | 129 | ||
| AL556162.3 | - | 3 - 4 | 278 | 129 | ||
| CX865814.1 | - | 5 - 6 | 631 | 129 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001459.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000482868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000482868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000482868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000482868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018074.hg.1 | TC09001459.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018074.hg.1 | ENST00000223608.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018074.hg.1 | ENST00000374586.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018074.hg.1 | ENST00000374587.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018074.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900175864.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175864.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175864.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175864.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900175864.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175864.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175864.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175864.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900175864.hg.1 | TC0900011151.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000482868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000482868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000482868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000482868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |

TMEM245
Genomic Map