Assay Details
| Gene Symbol: HCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK055840.1 | - | 7 - 8 | 2791 | 61 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000492035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000492035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000492035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000492035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000492035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000492035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |

HCN3
Genomic Map