| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Clariom D Assay |
PSR0600183669.hg.1 |
ENST00000312431.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183669.hg.1 |
ENST00000370754.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183669.hg.1 |
ENST00000449297.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183669.hg.1 |
ENST00000517937.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183669.hg.1 |
ENST00000520645.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183669.hg.1 |
ENST00000523967.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183669.hg.1 |
ENST00000524302.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183669.hg.1 |
NM_001144769.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600183669.hg.1 |
NM_001144770.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600183669.hg.1 |
ENST00000312431.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183669.hg.1 |
ENST00000370754.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183669.hg.1 |
ENST00000449297.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183669.hg.1 |
ENST00000517937.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183669.hg.1 |
ENST00000520645.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183669.hg.1 |
ENST00000523967.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183669.hg.1 |
ENST00000524302.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183669.hg.1 |
NM_001144769.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600183669.hg.1 |
NM_001144770.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600183669.hg.1 |
TC0600012123.hg.1 |
Clariom D TC |