Assay Details
| Gene Symbol: DST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011514826.2 | XP_011513128.1 | 96 - 97 | 22517 | 110 | |
| XM_017011210.1 | XP_016866699.1 | 101 - 102 | 23008 | 110 | ||
| XM_017011211.1 | XP_016866700.1 | 100 - 101 | 22936 | 110 | ||
| XM_017011215.1 | XP_016866704.1 | 100 - 101 | 22681 | 110 | ||
| XM_017011216.1 | XP_016866705.1 | 99 - 100 | 22609 | 110 | ||
| XM_017011224.1 | XP_016866713.1 | 91 - 92 | 16259 | 110 | ||
| GenBank mRNA | BC004912.2 | - | 3 - 4 | 395 | 110 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

DST
Genomic Map