Assay Details
Gene Symbol: RFX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022841.5 | NP_073752.5 | 3 - 4 | 280 | 65 | |
XM_005254603.3 | XP_005254660.2 | 4 - 5 | 374 | 65 | ||
XM_011521925.2 | XP_011520227.1 | 3 - 4 | 212 | 65 | ||
XM_017022506.1 | XP_016877995.1 | 4 - 5 | 3097 | 65 | ||
XM_017022507.1 | XP_016877996.1 | 3 - 4 | 340 | 65 | ||
XM_017022508.1 | XP_016877997.1 | 2 - 3 | 1509 | 65 | ||
GenBank mRNA | AK128045.1 | - | 3 - 4 | 217 | 65 | |
BC112936.1 | - | 1 - 2 | 100 | 6014850 | 65 | |
BC143438.1 | - | 1 - 2 | 37 | 65 | ||
DW009813.1 | - | 3 - 4 | 65 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC15001474.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK128045.1,BC112936.1,XM_011521925.2,XM_017022507.1,XM_017022508.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR15013856.hg.1 | ENST00000317318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15013856.hg.1 | ENST00000422057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15013856.hg.1 | ENST00000423270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15013856.hg.1 | ENST00000559447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15013856.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15013856.hg.1 | TC15001474.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15013854.hg.1 | ENST00000317318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15013854.hg.1 | ENST00000422057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15013854.hg.1 | ENST00000423270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15013854.hg.1 | ENST00000559447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15013854.hg.1 | ENST00000559847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15013854.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15013854.hg.1 | TC15001474.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR15013853.hg.1 | ENST00000317318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15013853.hg.1 | ENST00000422057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15013853.hg.1 | ENST00000423270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15013853.hg.1 | ENST00000559447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15013853.hg.1 | ENST00000559847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15013853.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15013853.hg.1 | TC15001474.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR15013858.hg.1 | ENST00000317318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15013858.hg.1 | ENST00000422057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15013858.hg.1 | ENST00000423270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15013858.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15013858.hg.1 | TC15001474.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR15013855.hg.1 | ENST00000317318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15013855.hg.1 | ENST00000422057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15013855.hg.1 | ENST00000423270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15013855.hg.1 | ENST00000559447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15013855.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15013855.hg.1 | TC15001474.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1500170247.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170247.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170247.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170247.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500170247.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170247.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170247.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170247.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500170247.hg.1 | TC1500009514.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500170255.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170255.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170255.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500170255.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170255.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170255.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500170255.hg.1 | TC1500009514.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500170245.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170245.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170245.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170245.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500170245.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170245.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170245.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170245.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500170245.hg.1 | TC1500009514.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500170249.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170249.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170249.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170249.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500170249.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170249.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170249.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170249.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500170249.hg.1 | TC1500009514.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500170258.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170258.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500170258.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170258.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500170258.hg.1 | TC1500009514.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500170250.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170250.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170250.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500170250.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170250.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500170250.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500170250.hg.1 | TC1500009514.hg.1 | Clariom D TC |
