Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013848.hg.1 |
ENST00000317318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013848.hg.1 |
ENST00000422057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013848.hg.1 |
ENST00000423270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013848.hg.1 |
ENST00000559447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013848.hg.1 |
ENST00000559847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013848.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013848.hg.1 |
TC15001474.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559847.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559847.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559847.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559847.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559847.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559847.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559847.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559847.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000559847.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000560792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000560792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000560792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000560792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000560792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000560792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000560792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
ENST00000560792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009514.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559847.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559847.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559847.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559847.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559847.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559847.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559847.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559847.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000559847.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000560792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000560792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000560792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000560792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000560792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000560792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000560792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
ENST00000560792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009514.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500170242.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500170242.hg.1 |
ENST00000559847.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500170242.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500170242.hg.1 |
ENST00000559447.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500170242.hg.1 |
ENST00000559847.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500170242.hg.1 |
NM_022841.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500170242.hg.1 |
TC1500009514.hg.1 |
Clariom D TC |