Assay Details
| Gene Symbol: COL6A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001848.2 | NP_001839.2 | 6 - 7 | 840 | 70 | |
| GenBank mRNA | AK298968.1 | - | 6 - 7 | 819 | 70 | |
| AK299396.1 | - | 6 - 7 | 822 | 70 | ||
| AK307989.1 | - | 6 - 7 | 819 | 70 | ||
| BC052575.1 | - | 6 - 7 | 820 | 6598940 | 70 | |
| BI769554.1 | - | 10 - 11 | 70 | |||
| X15879.1 | - | 6 - 7 | 70 | |||
More Information
Set Membership: |
|

COL6A1
Genomic Map