Assay Details
| Gene Symbol: COL6A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001848.2 | NP_001839.2 | 7 - 8 | 862 | 121 | |
| GenBank mRNA | AK298968.1 | - | 7 - 8 | 841 | 121 | |
| AK299396.1 | - | 7 - 8 | 844 | 121 | ||
| AK307989.1 | - | 7 - 8 | 841 | 121 | ||
| BC052575.1 | - | 7 - 8 | 842 | 6598940 | 121 | |
| BI769554.1 | - | 9 - 10 | 121 | |||
More Information
Updated Mapping Information
Set Membership: |
|

COL6A1
Genomic Map