Assay Details
| Gene Symbol: RRP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015056.2 | NP_055871.1 | 1 - 2 | 241 | 80 | |
| GenBank mRNA | AK056263.1 | - | 1 - 2 | 204 | 80 | |
| BC014005.2 | - | 1 - 2 | 213 | 80 | ||
| BC028386.2 | - | 1 - 2 | 246 | 4821672 | 80 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000212.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000470886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000470886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000470886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000470886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000470886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000470886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000470886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | ENST00000470886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002254.hg.1 | ENST00000340648.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002254.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002254.hg.1 | TC21000212.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002253.hg.1 | ENST00000340648.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002253.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002253.hg.1 | TC21000212.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2100149313.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100149313.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100149313.hg.1 | TC2100007318.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100149313.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100149313.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000470886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000470886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000470886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000470886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000470886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000470886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000470886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | ENST00000470886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007318.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100149314.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100149314.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100149314.hg.1 | TC2100007318.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100149314.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100149314.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100149315.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100149315.hg.1 | NM_015056.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100149315.hg.1 | TC2100007318.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100149315.hg.1 | ENST00000340648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100149315.hg.1 | NM_015056.2 | Refseq |

RRP1B
Genomic Map