Assay Details
| Gene Symbol: UPB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016327.2 | NP_057411.1 | 8 - 9 | 1041 | 73 | |
| XM_011530222.2 | XP_011528524.1 | 10 - 11 | 1345 | 73 | ||
| XM_011530225.2 | XP_011528527.1 | 9 - 10 | 1041 | 73 | ||
| GenBank mRNA | AB013885.1 | - | 8 - 9 | 980 | 73 | |
| AF163312.1 | - | 8 - 9 | 981 | 73 | ||
| AK024277.1 | - | 8 - 9 | 2309 | 73 | ||
| AK314727.1 | - | 8 - 9 | 1177 | 73 | ||
| BC033388.1 | - | 8 - 9 | 938 | 73 | ||
| BC131703.1 | - | 8 - 9 | 994 | 40121343 | 73 | |
| BC139843.1 | - | 8 - 9 | 939 | 73 | ||
| CR456375.1 | - | 8 - 9 | 1041 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000149.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22002818.hg.1 | ENST00000326010.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002818.hg.1 | ENST00000413389.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002818.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002818.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002818.hg.1 | TC22000149.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000382760.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000382760.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000382760.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000382760.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000382760.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000382760.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000382760.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000486043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000486043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000486043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000486043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002819.hg.1 | ENST00000326010.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002819.hg.1 | ENST00000413389.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002819.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002819.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002819.hg.1 | TC22000149.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000382760.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000382760.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000382760.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000382760.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000382760.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000382760.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000382760.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000486043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000486043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000486043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000486043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006894.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147758.hg.1 | TC2200006894.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200147758.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147758.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147758.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147758.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147758.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147758.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147758.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147758.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147757.hg.1 | TC2200006894.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200147757.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147757.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147757.hg.1 | ENST00000486043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147757.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147757.hg.1 | NM_016327.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147757.hg.1 | ENST00000326010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147757.hg.1 | ENST00000415388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147757.hg.1 | ENST00000486043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147757.hg.1 | ENST00000498140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147757.hg.1 | NM_016327.2 | Refseq |

UPB1
Genomic Map