Assay Details
| Gene Symbol: CYP27B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000785.3 | NP_000776.1 | 7 - 8 | 1371 | 76 | |
| GenBank mRNA | AB005038.1 | - | 7 - 8 | 1340 | 76 | |
| AB005989.1 | - | 7 - 8 | 1219 | 76 | ||
| AF020192.1 | - | 7 - 8 | 1231 | 76 | ||
| AF246895.1 | - | 7 - 8 | 1219 | 76 | ||
| AK314953.1 | - | 7 - 8 | 1255 | 76 | ||
| BC136386.1 | - | 7 - 8 | 1249 | 9020373 | 76 | |
| DQ925362.1 | - | 3 - 4 | 281 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001634.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
vitamin metabolic process
calcium ion transport
negative regulation of cell proliferation
negative regulation of calcidiol 1-monooxygenase activity
positive regulation of vitamin D 24-hydroxylase activity
bone mineralization
negative regulation of cell growth
regulation of bone mineralization
response to lipopolysaccharide
response to vitamin D
response to interferon-gamma
calcitriol biosynthetic process from calciol
vitamin D metabolic process
vitamin D catabolic process
response to estrogen
positive regulation of keratinocyte differentiation
decidualization
calcium ion homeostasis
oxidation-reduction process
G1 to G0 transition
positive regulation of vitamin D receptor signaling pathway
calcidiol 1-monooxygenase activity
iron ion binding
heme binding
vitamin metabolic process
calcium ion transport
negative regulation of cell proliferation
negative regulation of calcidiol 1-monooxygenase activity
positive regulation of vitamin D 24-hydroxylase activity
bone mineralization
negative regulation of cell growth
regulation of bone mineralization
response to lipopolysaccharide
response to vitamin D
response to interferon-gamma
calcitriol biosynthetic process from calciol
vitamin D metabolic process
vitamin D catabolic process
response to estrogen
positive regulation of keratinocyte differentiation
decidualization
calcium ion homeostasis
oxidation-reduction process
G1 to G0 transition
positive regulation of vitamin D receptor signaling pathway
calcidiol 1-monooxygenase activity
iron ion binding
heme binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000546496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000546496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000546567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000546567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000546567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000546567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000546567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000546567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000546567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000546567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000546567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000547451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000552186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000552186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000552186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | ENST00000552186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12022102.hg.1 | ENST00000228606.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12022102.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12022102.hg.1 | TC12001634.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200203927.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203927.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203927.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200203927.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203927.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203927.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200203927.hg.1 | TC1200012814.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200203926.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203926.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203926.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200203926.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203926.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203926.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200203926.hg.1 | TC1200012814.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000228606.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000546496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000546496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000546567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000546567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000546567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000546567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000546567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000546567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000546567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000546567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000546567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000547451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000552186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000552186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000552186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | ENST00000552186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | NM_000785.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012814.hg.1 | NM_000785.3 | Refseq |

CYP27B1
Genomic Map