| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22006052.hg.1 |
ENST00000336649.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22006052.hg.1 |
ENST00000400164.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22006052.hg.1 |
ENST00000401624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22006052.hg.1 |
ENST00000402142.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22006052.hg.1 |
ENST00000404898.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22006052.hg.1 |
ENST00000407673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22006052.hg.1 |
NM_001003406.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22006052.hg.1 |
NM_021096.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22006052.hg.1 |
TC22000318.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2200152279.hg.1 |
TC2200007407.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200152279.hg.1 |
ENST00000401624.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200152279.hg.1 |
ENST00000402142.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200152279.hg.1 |
ENST00000404898.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200152279.hg.1 |
ENST00000407673.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200152279.hg.1 |
NM_001003406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200152279.hg.1 |
NM_021096.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200152279.hg.1 |
ENST00000401624.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200152279.hg.1 |
ENST00000402142.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200152279.hg.1 |
ENST00000404898.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200152279.hg.1 |
ENST00000407673.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200152279.hg.1 |
NM_001003406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200152279.hg.1 |
NM_021096.3 |
Refseq |