Assay Details
| Gene Symbol: IKBIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153687.3 | NP_710154.1 | 1 - 2 | 529 | 159 | |
| NM_201612.2 | NP_963906.1 | 1 - 2 | 529 | 159 | ||
| GenBank mRNA | AJ539425.1 | - | 1 - 2 | 289 | 159 | |
| AJ539426.1 | - | 1 - 2 | 289 | 159 | ||
| AK055613.1 | - | 1 - 2 | 244 | 159 | ||
| BC058933.1 | - | 1 - 2 | 237 | 5541278 | 159 | |
| BM468651.1 | - | 1 - 2 | 529 | 159 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001861.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12024349.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024349.hg.1 | ENST00000342502.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024349.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024349.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024349.hg.1 | TC12001861.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024348.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024348.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024348.hg.1 | TC12001861.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024351.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024351.hg.1 | ENST00000342502.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024351.hg.1 | ENST00000393042.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024351.hg.1 | ENST00000420861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024351.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024351.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024351.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024351.hg.1 | TC12001861.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024350.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024350.hg.1 | ENST00000342502.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024350.hg.1 | ENST00000393042.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024350.hg.1 | ENST00000420861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024350.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024350.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024350.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024350.hg.1 | TC12001861.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | ENST00000393042.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | ENST00000393042.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | ENST00000393042.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | ENST00000393042.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | ENST00000393042.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | ENST00000393042.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011656.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024345.hg.1 | ENST00000342502.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024345.hg.1 | ENST00000393042.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024345.hg.1 | ENST00000420861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024345.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024345.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024345.hg.1 | TC12001861.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200190191.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190191.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190191.hg.1 | ENST00000393042.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190191.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190191.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190191.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190191.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190191.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190191.hg.1 | ENST00000393042.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190191.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190191.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190191.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190191.hg.1 | TC1200011656.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200190192.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190192.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190192.hg.1 | ENST00000393042.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190192.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190192.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190192.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190192.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190192.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190192.hg.1 | ENST00000393042.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190192.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190192.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190192.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190192.hg.1 | TC1200011656.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200190190.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190190.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190190.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190190.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190190.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190190.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190190.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190190.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190190.hg.1 | TC1200011656.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200190194.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190194.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190194.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190194.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190194.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190194.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190194.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190194.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190194.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190194.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190194.hg.1 | TC1200011656.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200190193.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190193.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190193.hg.1 | ENST00000393042.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190193.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190193.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190193.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190193.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190193.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190193.hg.1 | ENST00000393042.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200190193.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190193.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190193.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200190193.hg.1 | TC1200011656.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | ENST00000299157.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | ENST00000342502.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | ENST00000393042.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | ENST00000393042.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | ENST00000393042.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | ENST00000393042.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | ENST00000393042.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | ENST00000393042.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | NM_153687.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | NM_201612.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | NM_201613.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011656.hg.1 | NM_201613.2 | Refseq |

IKBIP
Genomic Map