Assay Details
| Gene Symbol: PRIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000946.2 | NP_000937.1 | 11 - 12 | 1201 | 115 | |
| GenBank mRNA | AA761682.1 | - | 2 - 3 | 115 | ||
| AK223391.1 | - | 11 - 12 | 1162 | 115 | ||
| AK314866.1 | - | 11 - 12 | 1162 | 115 | ||
| BC005266.1 | - | 11 - 12 | 1174 | 3686937 | 115 | |
| X74330.1 | - | 11 - 12 | 1162 | 115 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001614.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12021644.hg.1 | ENST00000338193.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021644.hg.1 | ENST00000537418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021644.hg.1 | ENST00000552853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021644.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021644.hg.1 | TC12001614.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000546761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000546761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000546761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000546761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000549549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000549549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000549549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000549549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000549549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200203634.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203634.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203634.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200203634.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203634.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203634.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200203634.hg.1 | TC1200012804.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200203635.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203635.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203635.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200203635.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203635.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203635.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200203635.hg.1 | TC1200012804.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000546761.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000546761.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000546761.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000546761.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000549549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000549549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000549549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000549549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000549549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |

PRIM1
Genomic Map