Assay Details
Gene Symbol: PRIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000946.2 | NP_000937.1 | 6 - 7 | 701 | 165 | |
GenBank mRNA | AK223391.1 | - | 6 - 7 | 662 | 165 | |
AK314866.1 | - | 6 - 7 | 662 | 165 | ||
BC005266.1 | - | 6 - 7 | 674 | 3686937 | 165 | |
X74330.1 | - | 6 - 7 | 662 | 165 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001614.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12021652.hg.1 | ENST00000338193.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12021652.hg.1 | ENST00000537418.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12021652.hg.1 | ENST00000552853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12021652.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12021652.hg.1 | TC12001614.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000546761.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000546761.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000546761.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000546761.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000549549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000549549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000549549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000549549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000549549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552408.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552408.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552408.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000546761.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000546761.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000546761.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000546761.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000548173.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000549549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000549549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000549549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000549549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000549549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552408.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552408.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552408.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012804.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200203642.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200203642.hg.1 | ENST00000549549.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200203642.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200203642.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200203642.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200203642.hg.1 | ENST00000338193.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200203642.hg.1 | ENST00000549549.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200203642.hg.1 | ENST00000550770.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200203642.hg.1 | ENST00000552590.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200203642.hg.1 | NM_000946.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200203642.hg.1 | TC1200012804.hg.1 | Clariom D TC |
