Assay Details
| Gene Symbol: EPHA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080448.2 | NP_001073917.2 | 6 - 7 | 1777 | 76 | |
| XM_017006221.1 | XP_016861710.1 | 1 - 2 | 135 | 76 | ||
| GenBank mRNA | DV080244.1 | - | 1 - 2 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017006221.1 transcripts.
Set Membership: |
|

EPHA6
Genomic Map