Assay Details
| Gene Symbol: EPHA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080448.2 | NP_001073917.2 | 7 - 8 | 1932 | 74 | |
| XM_006713592.3 | XP_006713655.1 | 8 - 9 | 1980 | 74 | ||
| XM_017006210.1 | XP_016861699.1 | 8 - 9 | 1980 | 74 | ||
| XM_017006211.1 | XP_016861700.1 | 9 - 10 | 2005 | 74 | ||
| XM_017006212.1 | XP_016861701.1 | 8 - 9 | 1674 | 74 | ||
| XM_017006213.1 | XP_016861702.1 | 8 - 9 | 2032 | 74 | ||
| XM_017006214.1 | XP_016861703.1 | 7 - 8 | 1300 | 74 | ||
| XM_017006215.1 | XP_016861704.1 | 3 - 4 | 329 | 74 | ||
| XM_017006216.1 | XP_016861705.1 | 2 - 3 | 279 | 74 | ||
| XM_017006217.1 | XP_016861706.1 | 4 - 5 | 376 | 74 | ||
| XM_017006218.1 | XP_016861707.1 | 2 - 3 | 179 | 74 | ||
| XM_017006221.1 | XP_016861710.1 | 2 - 3 | 290 | 74 | ||
| XM_017006222.1 | XP_016861711.1 | 3 - 4 | 338 | 74 | ||
| GenBank mRNA | AK302080.1 | - | 3 - 4 | 234 | 74 | |
| AK310249.1 | - | 3 - 4 | 263 | 74 | ||
| DC398430.1 | - | 3 - 4 | 282 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK302080.1, XM_017006221.1, XM_017006222.1 transcripts.
Set Membership: |
|

EPHA6
Genomic Map