Assay Details
| Gene Symbol: AIFM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198696.1 | NP_001185625.1 | 2 - 3 | 396 | 97 | |
| NM_032797.5 | NP_116186.1 | 2 - 3 | 393 | 97 | ||
| GenBank mRNA | AF337957.1 | - | 1 - 2 | 197 | 97 | |
| AF506757.1 | - | 2 - 3 | 393 | 97 | ||
| AK027403.1 | - | 2 - 3 | 272 | 97 | ||
| AK127353.1 | - | 2 - 3 | 2148 | 97 | ||
| BC006121.1 | - | 1 - 2 | 201 | 3506309 | 97 | |
| BC023601.2 | - | 2 - 3 | 380 | 4548458 | 97 | |
| BG332787.1 | - | 2 - 3 | 381 | 97 | ||
| BP331371.1 | - | 2 - 3 | 312 | 97 | ||
| BX537621.1 | - | 1 - 2 | 113 | 97 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001359.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10017172.hg.1 | ENST00000307864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017172.hg.1 | ENST00000373248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017172.hg.1 | ENST00000395039.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017172.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017172.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017172.hg.1 | TC10001359.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000482166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000482166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000482166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000482166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000482166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017173.hg.1 | ENST00000307864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017173.hg.1 | ENST00000373248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017173.hg.1 | ENST00000395039.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017173.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017173.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017173.hg.1 | TC10001359.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017179.hg.1 | ENST00000307864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017179.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017179.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017179.hg.1 | TC10001359.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017176.hg.1 | ENST00000307864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017176.hg.1 | ENST00000395039.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017176.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017176.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017176.hg.1 | TC10001359.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017178.hg.1 | ENST00000307864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017178.hg.1 | ENST00000395039.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017178.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017178.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017178.hg.1 | TC10001359.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000482166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000482166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000482166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000482166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000482166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010909.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000174234.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000174234.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000174234.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000174234.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000174234.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000174234.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000174234.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000174234.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000174234.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000174234.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000174234.hg.1 | TC1000010909.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000174239.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000174239.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000174239.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000174239.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000174239.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000174239.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000174239.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000174239.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000174239.hg.1 | TC1000010909.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000174235.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000174235.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000174235.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000174235.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000174235.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000174235.hg.1 | ENST00000307864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000174235.hg.1 | ENST00000373248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000174235.hg.1 | ENST00000613322.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000174235.hg.1 | NM_001198696.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000174235.hg.1 | NM_032797.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000174235.hg.1 | TC1000010909.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 129590 | - | - |

AIFM2
Genomic Map