| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01057252.hg.1 |
TC01003729.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01057252.hg.1 |
ENST00000272203.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01057252.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01057252.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01057253.hg.1 |
TC01003729.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01057253.hg.1 |
ENST00000272203.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01057253.hg.1 |
ENST00000367191.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01057253.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01057253.hg.1 |
ENST00000430806.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01057253.hg.1 |
ENST00000450705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01057253.hg.1 |
ENST00000454206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01057253.hg.1 |
ENST00000538441.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01057253.hg.1 |
ENST00000543129.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01057253.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000485632.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000485632.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000485632.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100244494.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100244494.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100244494.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100244494.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100244494.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100244494.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100244494.hg.1 |
TC0100018553.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000485632.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000485632.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000485632.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100244493.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100244493.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100244493.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100244493.hg.1 |
ENST00000272203.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100244493.hg.1 |
ENST00000414478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100244493.hg.1 |
NM_014935.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100244493.hg.1 |
TC0100018553.hg.1 |
Clariom D TC |