Assay Details
| Gene Symbol: PLEKHA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011509297.2 | XP_011507599.1 | 2 - 3 | 867 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLEKHA6| Gene Symbol: PLEKHA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011509297.2 | XP_011507599.1 | 2 - 3 | 867 | 103 | |
Set Membership: |
|