Assay Details
| Gene Symbol: PLEKHA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014935.4 | NP_055750.2 | 6 - 7 | 715 | 72 | |
| XM_005244966.4 | XP_005245023.2 | 6 - 7 | 582 | 72 | ||
| XM_005244968.4 | XP_005245025.2 | 6 - 7 | 581 | 72 | ||
| XM_017000689.1 | XP_016856178.1 | 6 - 7 | 582 | 72 | ||
| XM_017000690.1 | XP_016856179.1 | 6 - 7 | 4587 | 72 | ||
| GenBank mRNA | AB023186.1 | - | 6 - 7 | 702 | 72 | |
| BC152475.1 | - | 6 - 7 | 702 | 8862612 | 72 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003729.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000485632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000485632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000485632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057263.hg.1 | TC01003729.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057263.hg.1 | ENST00000272203.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057263.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057263.hg.1 | ENST00000485632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057263.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244505.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244505.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244505.hg.1 | ENST00000485632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244505.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244505.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244505.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244505.hg.1 | ENST00000485632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244505.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244505.hg.1 | TC0100018553.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000485632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000485632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000485632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |

PLEKHA6
Genomic Map