Assay Details
| Gene Symbol: PLEKHA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014935.4 | NP_055750.2 | 1 - 2 | 241 | 81 | |
| XM_006711221.2 | XP_006711284.1 | 1 - 2 | 432 | 81 | ||
| GenBank mRNA | AB023186.1 | - | 1 - 2 | 228 | 81 | |
| BC152475.1 | - | 1 - 2 | 228 | 8862612 | 81 | |
| BX954305.1 | - | 1 - 2 | 242 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003729.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB023186.1,BC152475.1,NM_014935.4,XM_006711221.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000485632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000485632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | ENST00000485632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057272.hg.1 | TC01003729.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057272.hg.1 | ENST00000272203.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057272.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057272.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057271.hg.1 | TC01003729.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057271.hg.1 | ENST00000272203.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057271.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057271.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244514.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244514.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244514.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244514.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244514.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244514.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244514.hg.1 | TC0100018553.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000485632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000485632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | ENST00000485632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018553.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244515.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244515.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244515.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244515.hg.1 | ENST00000272203.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244515.hg.1 | ENST00000414478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244515.hg.1 | NM_014935.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244515.hg.1 | TC0100018553.hg.1 | Clariom D TC |

PLEKHA6
Genomic Map