Assay Details
| Gene Symbol: IFI27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001288952.1 | NP_001275881.1 | 2 - 3 | 121 | 124 | |
| NM_001288954.1 | NP_001275883.1 | 2 - 3 | 121 | 124 | ||
| GenBank mRNA | BF971643.1 | - | 2 - 3 | 100 | 124 | |
| BP317624.1 | - | 2 - 3 | 118 | 124 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001288952.1, NM_001288954.1 transcripts.
Set Membership: |
|

IFI27
Genomic Map