Assay Details
| Gene Symbol: TAGAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054114.4 | NP_473455.2 | 3 - 4 | 407 | 119 | |
| NM_138810.3 | NP_620165.1 | 3 - 4 | 407 | 119 | ||
| NM_152133.2 | NP_687034.1 | 3 - 4 | 407 | 119 | ||
| GenBank mRNA | AF385429.1 | - | 3 - 4 | 407 | 119 | |
| AF385430.1 | - | 3 - 4 | 407 | 119 | ||
| AK097090.1 | - | 3 - 4 | 273 | 119 | ||
| AL832662.1 | - | 3 - 4 | 283 | 119 | ||
| BC015859.1 | - | 3 - 4 | 277 | 119 | ||
| BC111731.1 | - | 3 - 4 | 263 | 40035199 | 119 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06002270.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF385430.1,NM_152133.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06028493.hg.1 | TC06002270.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028493.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028493.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028493.hg.1 | ENST00000367066.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028493.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028493.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028493.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194721.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194721.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194721.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194721.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194721.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194721.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194721.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194721.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194721.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194721.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194721.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194721.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194721.hg.1 | TC0600013743.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000326965.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000338313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | ENST00000367066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_001278733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_054114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_138810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013743.hg.1 | NM_152133.2 | Refseq |

TAGAP
Genomic Map