Assay Details
| Gene Symbol: DGKE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003647.2 | NP_003638.1 | 9 - 10 | 1461 | 139 | |
| XM_011525394.2 | XP_011523696.1 | 9 - 10 | 1439 | 139 | ||
| XM_011525395.2 | XP_011523697.1 | 9 - 10 | 1446 | 139 | ||
| XM_017025243.1 | XP_016880732.1 | 9 - 10 | 1828 | 139 | ||
| GenBank mRNA | AB209365.1 | - | 8 - 9 | 1111 | 139 | |
| AK293039.1 | - | 9 - 10 | 1408 | 139 | ||
| BC130629.1 | - | 8 - 9 | 1305 | 40146660 | 139 | |
| BC130631.1 | - | 8 - 9 | 1305 | 40146662 | 139 | |
| U49379.1 | - | 9 - 10 | 1368 | 139 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000699.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17009157.hg.1 | ENST00000284061.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009157.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009157.hg.1 | TC17000699.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000570738.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000570738.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000570738.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000571084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000571084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000571084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000572810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000572810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000572810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000572810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000576602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000576869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000576869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000576869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000576869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000576869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000576869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | ENST00000576869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162309.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162309.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162309.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162309.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162309.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162309.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162309.hg.1 | TC1700008357.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000284061.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000570738.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000570738.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000570738.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000571084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000571084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000571084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000572810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000572810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000572810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000572810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000572944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000576602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000576869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000576869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000576869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000576869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000576869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000576869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | ENST00000576869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | NM_003647.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008357.hg.1 | NM_003647.2 | Refseq |

DGKE
Genomic Map