Assay Details
| Gene Symbol: ASCC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006828.3 | NP_006819.2 | 32 - 33 | 5396 | 119 | |
| XM_011535394.2 | XP_011533696.1 | 32 - 33 | 5406 | 119 | ||
| XM_011535395.2 | XP_011533697.1 | 33 - 34 | 5631 | 119 | ||
| XM_011535396.2 | XP_011533698.1 | 31 - 32 | 5240 | 119 | ||
| XM_017010205.1 | XP_016865694.1 | 33 - 34 | 5652 | 119 | ||
| XM_017010206.1 | XP_016865695.1 | 33 - 34 | 5430 | 119 | ||
| GenBank mRNA | AJ223948.1 | - | 15 - 16 | 2235 | 119 | |
| AL834463.1 | - | 31 - 32 | 5081 | 119 | ||
| AY013288.1 | - | 28 - 29 | 4215 | 119 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001970.hg.1
Set Membership: |
|

ASCC3
Genomic Map