Assay Details
| Gene Symbol: KCNG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006723785.2 | XP_006723848.1 | 3 - 4 | 650 | 64 | |
| XM_011528800.1 | XP_011527102.1 | 2 - 3 | 325 | 64 | ||
| XM_011528801.1 | XP_011527103.1 | 3 - 4 | 434 | 64 | ||
| XM_011528802.1 | XP_011527104.1 | 3 - 4 | 429 | 64 | ||
| XM_011528803.2 | XP_011527105.1 | 3 - 4 | 356 | 64 | ||
| XM_011528804.1 | XP_011527106.1 | 2 - 3 | 579 | 64 | ||
| GenBank mRNA | BC006367.1 | - | 2 - 3 | 483 | 4131541 | 64 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC006367.1,XM_006723785.2,XM_011528800.1,XM_011528801.1,XM_011528802.1,XM_011528803.2,XM_011528804.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000007718.hg.1 | ENST00000424566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007718.hg.1 | ENST00000424566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007718.hg.1 | ENST00000424566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000424171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000424171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000424171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000424171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000424171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000424171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000424171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000433903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000433903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000433903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000433903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000433903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000433903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000439216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000439216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000439216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000439216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000439216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000439216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000439216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000447736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000447736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000447736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000447736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000447736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000506387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000506387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000506387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | ENST00000506387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | NM_002237.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | NM_002237.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | NM_002237.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | NM_002237.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | NM_002237.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | NM_002237.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | NM_002237.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009448.hg.1 | NM_002237.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000166085.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166085.hg.1 | ENST00000424171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166085.hg.1 | ENST00000433903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166085.hg.1 | ENST00000439216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166085.hg.1 | NM_002237.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000166085.hg.1 | TC2000009448.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000166085.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166085.hg.1 | ENST00000424171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166085.hg.1 | ENST00000433903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166085.hg.1 | ENST00000439216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166085.hg.1 | NM_002237.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000166086.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166086.hg.1 | ENST00000424171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166086.hg.1 | ENST00000433903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166086.hg.1 | ENST00000439216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166086.hg.1 | ENST00000447736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166086.hg.1 | NM_002237.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000166086.hg.1 | TC2000009448.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000166086.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166086.hg.1 | ENST00000424171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166086.hg.1 | ENST00000433903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166086.hg.1 | ENST00000439216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166086.hg.1 | ENST00000447736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166086.hg.1 | NM_002237.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000166087.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166087.hg.1 | ENST00000424171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166087.hg.1 | ENST00000433903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166087.hg.1 | ENST00000439216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166087.hg.1 | ENST00000447736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166087.hg.1 | NM_002237.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000166087.hg.1 | TC2000009448.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000166087.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166087.hg.1 | ENST00000424171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166087.hg.1 | ENST00000433903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166087.hg.1 | ENST00000439216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166087.hg.1 | ENST00000447736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166087.hg.1 | NM_002237.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000166082.hg.1 | TC2000009448.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000166084.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166084.hg.1 | ENST00000424171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166084.hg.1 | ENST00000439216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166084.hg.1 | NM_002237.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000166084.hg.1 | TC2000009448.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000166084.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166084.hg.1 | ENST00000424171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166084.hg.1 | ENST00000439216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166084.hg.1 | NM_002237.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007718.hg.1 | ENST00000424566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007718.hg.1 | ENST00000424566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007718.hg.1 | ENST00000424566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000371571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000424171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000424171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000424171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000424171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000424171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000424171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000424171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000433903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000433903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000433903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000433903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000433903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000433903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000439216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000439216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000439216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000439216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000439216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000439216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000439216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000447736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000447736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000447736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000447736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000447736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000506387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000506387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000506387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | ENST00000506387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | NM_002237.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | NM_002237.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | NM_002237.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | NM_002237.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | NM_002237.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | NM_002237.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | NM_002237.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009448.hg.1 | NM_002237.3 | Refseq |

KCNG1
Genomic Map