| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039583.hg.1 |
TC01002544.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039583.hg.1 |
ENST00000235628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039583.hg.1 |
NM_032526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013801.hg.1 |
ENST00000235628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013801.hg.1 |
ENST00000235628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013801.hg.1 |
ENST00000235628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013801.hg.1 |
ENST00000235628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013801.hg.1 |
ENST00000235628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013801.hg.1 |
ENST00000235628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013801.hg.1 |
NM_032526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013801.hg.1 |
NM_032526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013801.hg.1 |
NM_032526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013801.hg.1 |
NM_032526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013801.hg.1 |
NM_032526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013801.hg.1 |
NM_032526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100203206.hg.1 |
ENST00000235628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100203206.hg.1 |
NM_032526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100203206.hg.1 |
ENST00000235628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100203206.hg.1 |
NM_032526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100203206.hg.1 |
TC0100013801.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100013801.hg.1 |
ENST00000235628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013801.hg.1 |
ENST00000235628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013801.hg.1 |
ENST00000235628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013801.hg.1 |
ENST00000235628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013801.hg.1 |
ENST00000235628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013801.hg.1 |
ENST00000235628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013801.hg.1 |
NM_032526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013801.hg.1 |
NM_032526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013801.hg.1 |
NM_032526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013801.hg.1 |
NM_032526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013801.hg.1 |
NM_032526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013801.hg.1 |
NM_032526.2 |
Refseq |