Assay Details
| Gene Symbol: NPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000906.3 | NP_000897.3 | 6 - 7 | 1815 | 82 | |
| XM_005245218.1 | XP_005245275.1 | 6 - 7 | 1886 | 82 | ||
| XM_017001374.1 | XP_016856863.1 | 6 - 7 | 1886 | 82 | ||
| GenBank mRNA | AK025024.1 | - | 6 - 7 | 1815 | 82 | |
| AK300446.1 | - | 5 - 6 | 1021 | 82 | ||
| BC063304.1 | - | 6 - 7 | 1864 | 30331757 | 82 | |
| X15357.1 | - | 6 - 7 | 1437 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001262.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cGMP biosynthetic process
protein phosphorylation
cell surface receptor signaling pathway
receptor guanylyl cyclase signaling pathway
G-protein coupled receptor signaling pathway
body fluid secretion
regulation of blood pressure
negative regulation of angiogenesis
negative regulation of cell growth
positive regulation of cGMP biosynthetic process
intracellular signal transduction
positive regulation of urine volume
positive regulation of renal sodium excretion
regulation of vasodilation
dopamine metabolic process
regulation of vascular permeability
negative regulation of smooth muscle cell proliferation
regulation of cardiac conduction
adenylate cyclase activity
guanylate cyclase activity
protein kinase activity
ATP binding
GTP binding
G-protein coupled peptide receptor activity
natriuretic peptide receptor activity
peptide hormone binding
protein kinase binding
hormone binding
cGMP biosynthetic process
protein phosphorylation
cell surface receptor signaling pathway
receptor guanylyl cyclase signaling pathway
G-protein coupled receptor signaling pathway
body fluid secretion
regulation of blood pressure
negative regulation of angiogenesis
negative regulation of cell growth
positive regulation of cGMP biosynthetic process
intracellular signal transduction
positive regulation of urine volume
positive regulation of renal sodium excretion
regulation of vasodilation
dopamine metabolic process
regulation of vascular permeability
negative regulation of smooth muscle cell proliferation
regulation of cardiac conduction
adenylate cyclase activity
guanylate cyclase activity
protein kinase activity
ATP binding
GTP binding
G-protein coupled peptide receptor activity
natriuretic peptide receptor activity
peptide hormone binding
protein kinase binding
hormone binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368677.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368677.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368677.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368677.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368677.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368677.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368677.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368677.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368677.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368677.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000413826.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000413826.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | ENST00000413826.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019245.hg.1 | TC01001262.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019245.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019245.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172812.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172812.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172812.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172812.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172812.hg.1 | TC0100010047.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368677.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368677.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368677.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368677.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368677.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368677.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368677.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368677.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368677.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368677.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000368680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000413826.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000413826.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | ENST00000413826.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010047.hg.1 | NM_000906.3 | Refseq |

NPR1
Genomic Map