Assay Details
| Gene Symbol: RRAGB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016656.3 | NP_057740.2 | 3 - 4 | 873 | 145 | |
| XM_005261973.4 | XP_005262030.1 | 3 - 4 | 210 | 145 | ||
| XM_017029218.1 | XP_016884707.1 | 3 - 4 | 829 | 145 | ||
| GenBank mRNA | X90530.1 | - | 3 - 4 | 664 | 145 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000335.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cell cycle arrest
regulation of autophagy
macroautophagy
regulation of TOR signaling
positive regulation of TOR signaling
cellular response to amino acid starvation
cellular protein localization
cellular response to amino acid stimulus
GTPase activity
protein binding
GTP binding
guanyl ribonucleotide binding
cell cycle arrest
regulation of autophagy
macroautophagy
regulation of TOR signaling
positive regulation of TOR signaling
cellular response to amino acid starvation
cellular protein localization
cellular response to amino acid stimulus
GTPase activity
protein binding
GTP binding
guanyl ribonucleotide binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000414239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000414239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000414239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000414239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000414239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000414239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000414239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000414239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000474757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000474757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000474757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000475061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000475061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000475061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000475061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000475061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000498762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000498762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000498762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000498762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | ENST00000498762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004417.hg.1 | TC0X000335.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004417.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004417.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150776.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150776.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150776.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150776.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150776.hg.1 | TC0X00007382.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000262850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000374941.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000414239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000414239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000414239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000414239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000414239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000414239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000414239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000414239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000474757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000474757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000474757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000475061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000475061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000475061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000475061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000475061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000498762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000498762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000498762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000498762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | ENST00000498762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_006064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_016656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007382.hg.1 | NM_016656.3 | Refseq |

RRAGB
Genomic Map