Assay Details
| Gene Symbol: DLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023932.3 | NP_076421.2 | 4 - 5 | 552 | 80 | |
| NM_206539.2 | NP_996262.1 | 4 - 5 | 968 | 80 | ||
| XM_005249308.4 | XP_005249365.1 | 3 - 4 | 1760 | 80 | ||
| XM_017011204.1 | XP_016866693.1 | 1 - 2 | 570 | 80 | ||
| GenBank mRNA | AK055380.1 | - | 4 - 5 | 968 | 80 | |
| AY358126.1 | - | 4 - 5 | 418 | 80 | ||
| BC000230.1 | - | 4 - 5 | 384 | 3350499 | 80 | |
| BC006425.1 | - | 4 - 5 | 384 | 3349698 | 80 | |
| BC110320.1 | - | 4 - 5 | 520 | 5495458 | 80 | |
| BI916137.1 | - | 4 - 5 | 476 | 80 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC000230.1,BC006425.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000430324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000430324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000430324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000430324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | ENST00000430324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_001286655.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_001286655.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_001286655.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_001286655.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_001286655.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_001286655.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_001286655.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_001286655.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_001286655.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_001286655.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_001286655.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011874.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181532.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181532.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181532.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181532.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181532.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181532.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181532.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181532.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181532.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181532.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181532.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181532.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181532.hg.1 | TC0600011874.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000357338.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000372485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000372488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000430324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000430324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000430324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000430324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | ENST00000430324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_001286655.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_001286655.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_001286655.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_001286655.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_001286655.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_001286655.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_001286655.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_001286655.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_001286655.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_001286655.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_001286655.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_001286656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_023932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_206539.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011874.hg.1 | NM_206539.2 | Refseq |

DLK2
Genomic Map