Assay Details
| Gene Symbol: PORCN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203475.2 | NP_982301.1 | 6 - 7 | 870 | 76 | |
| XM_005272637.1 | XP_005272694.1 | 7 - 8 | 1152 | 76 | ||
| XM_006724545.3 | XP_006724608.1 | 7 - 8 | 935 | 76 | ||
| XM_006724546.3 | XP_006724609.1 | 7 - 8 | 1007 | 76 | ||
| XM_006724547.1 | XP_006724610.1 | 6 - 7 | 727 | 76 | ||
| XM_017029735.1 | XP_016885224.1 | 6 - 7 | 1235 | 76 | ||
| GenBank mRNA | AF317058.1 | - | 5 - 6 | 691 | 76 | |
| AK314745.1 | - | 5 - 6 | 740 | 76 | ||
| L08239.1 | - | 4 - 5 | 607 | 76 | ||
More Information
Set Membership: |
|

PORCN
Genomic Map