Assay Details
| Gene Symbol: GOPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017408.2 | NP_001017408.1 | 1 - 2 | 540 | 81 | |
| NM_020399.3 | NP_065132.1 | 1 - 2 | 540 | 81 | ||
| GenBank mRNA | AB209385.1 | - | 1 - 2 | 486 | 81 | |
| AF287894.1 | - | 1 - 2 | 362 | 81 | ||
| AF450008.1 | - | 1 - 2 | 528 | 81 | ||
| AK314486.1 | - | 1 - 2 | 511 | 81 | ||
| AY033606.1 | - | 1 - 2 | 493 | 81 | ||
| BC009553.2 | - | 1 - 2 | 475 | 3898495 | 81 | |
| BX419592.2 | - | 1 - 2 | 475 | 81 | ||
| DA109449.1 | - | 1 - 2 | 541 | 81 | ||
| KF420122.1 | - | 1 - 2 | 286 | 81 | ||
| KF420123.1 | - | 1 - 2 | 286 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06004056.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06025690.hg.1 | TC06004056.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025690.hg.1 | ENST00000052569.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025690.hg.1 | ENST00000368498.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025690.hg.1 | ENST00000535237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025690.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025690.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025692.hg.1 | TC06004056.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025692.hg.1 | ENST00000052569.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025692.hg.1 | ENST00000535237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025692.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025692.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025691.hg.1 | TC06004056.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025691.hg.1 | ENST00000052569.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025691.hg.1 | ENST00000368498.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025691.hg.1 | ENST00000535237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025691.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025691.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025693.hg.1 | TC06004056.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025693.hg.1 | ENST00000052569.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025693.hg.1 | ENST00000535237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025693.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025693.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202591.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202591.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202591.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202591.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202591.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202591.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202591.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202591.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202591.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202591.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202591.hg.1 | TC0600014337.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202592.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202592.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202592.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202592.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202592.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202592.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202592.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202592.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202592.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202592.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202592.hg.1 | TC0600014337.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600202594.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202594.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202594.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202594.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202594.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202594.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202594.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202594.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202594.hg.1 | TC0600014337.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600202593.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202593.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202593.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202593.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202593.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202593.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202593.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202593.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202593.hg.1 | TC0600014337.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600202590.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202590.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202590.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202590.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202590.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202590.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202590.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202590.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202590.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202590.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202590.hg.1 | TC0600014337.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 103651 | - | - |

GOPC
Genomic Map