Assay Details
| Gene Symbol: GOPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020399.3 | NP_065132.1 | 2 - 3 | 705 | 76 | |
| GenBank mRNA | AB209385.1 | - | 2 - 3 | 651 | 76 | |
| AF287894.1 | - | 2 - 3 | 527 | 76 | ||
| BX419592.2 | - | 2 - 3 | 640 | 76 | ||
| KF420123.1 | - | 2 - 3 | 451 | 76 | ||
More Information
Set Membership: |
|

GOPC
Genomic Map