Assay Details
| Gene Symbol: GOPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017408.2 | NP_001017408.1 | 7 - 8 | 1474 | 77 | |
| NM_020399.3 | NP_065132.1 | 8 - 9 | 1498 | 77 | ||
| GenBank mRNA | AB209385.1 | - | 7 - 8 | 2567 | 77 | |
| AF287894.1 | - | 8 - 9 | 1320 | 77 | ||
| AF450008.1 | - | 7 - 8 | 1462 | 77 | ||
| AK314486.1 | - | 7 - 8 | 1445 | 77 | ||
| AY033606.1 | - | 7 - 8 | 1427 | 77 | ||
| BC009553.2 | - | 7 - 8 | 1409 | 3898495 | 77 | |
| KF420122.1 | - | 6 - 7 | 1052 | 77 | ||
| KF420123.1 | - | 7 - 8 | 1079 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06004056.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06025674.hg.1 | TC06004056.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025674.hg.1 | ENST00000052569.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025674.hg.1 | ENST00000368498.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025674.hg.1 | ENST00000535237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025674.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025674.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025675.hg.1 | TC06004056.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025675.hg.1 | ENST00000052569.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025675.hg.1 | ENST00000368498.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025675.hg.1 | ENST00000535237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025675.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025675.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025676.hg.1 | TC06004056.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025676.hg.1 | ENST00000052569.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025676.hg.1 | ENST00000368498.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025676.hg.1 | ENST00000535237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025676.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025676.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014337.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202579.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202579.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202579.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202579.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202579.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202579.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202579.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202579.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202579.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202579.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202579.hg.1 | TC0600014337.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600202580.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202580.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202580.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202580.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202580.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202580.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202580.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202580.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202580.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202580.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202580.hg.1 | TC0600014337.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600202581.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202581.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202581.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202581.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202581.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202581.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202581.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202581.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202581.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202581.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202581.hg.1 | TC0600014337.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600202582.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202582.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202582.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202582.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202582.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202582.hg.1 | ENST00000052569.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202582.hg.1 | ENST00000368498.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202582.hg.1 | ENST00000535237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202582.hg.1 | NM_001017408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202582.hg.1 | NM_020399.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202582.hg.1 | TC0600014337.hg.1 | Clariom D TC |

GOPC
Genomic Map