Assay Details
| Gene Symbol: STX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001326580.1 | NP_001313509.1 | 1 - 2 | 193 | 72 | |
| NM_003569.2 | NP_003560.2 | 1 - 2 | 193 | 72 | ||
| NR_137169.1 | 1 - 2 | 193 | 72 | |||
| XM_011536178.1 | XP_011534480.1 | 1 - 2 | 109 | 72 | ||
| GenBank mRNA | AK294911.1 | - | 1 - 2 | 77 | 72 | |
| AK301464.1 | - | 1 - 2 | 81 | 72 | ||
| AW500305.1 | - | 1 - 2 | 196 | 72 | ||
| BC011975.1 | - | 1 - 2 | 57 | 3860059 | 72 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06002114.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK294911.1,AK301464.1,BC011975.1,NM_001326580.1,NM_003569.2,XM_011536178.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of T cell mediated cytotoxicity
intracellular protein transport
vesicle fusion
vesicle docking
organelle localization
organelle assembly
positive regulation of receptor localization to synapse
regulation of protein localization to plasma membrane
SNARE binding
SNAP receptor activity
protein binding
chloride channel inhibitor activity
syntaxin binding
positive regulation of T cell mediated cytotoxicity
intracellular protein transport
vesicle fusion
vesicle docking
organelle localization
organelle assembly
positive regulation of receptor localization to synapse
regulation of protein localization to plasma membrane
SNARE binding
SNAP receptor activity
protein binding
chloride channel inhibitor activity
syntaxin binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06026544.hg.1 | TC06002114.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026544.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026544.hg.1 | ENST00000367941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026544.hg.1 | ENST00000448348.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026544.hg.1 | ENST00000475879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026544.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026546.hg.1 | TC06002114.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026546.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026546.hg.1 | ENST00000448348.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026546.hg.1 | ENST00000475879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026546.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026548.hg.1 | TC06002114.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026548.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026548.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000448348.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000448348.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000448348.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000448348.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000448348.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000448348.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000448348.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000475879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000475879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000475879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000475879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | ENST00000475879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191100.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191100.hg.1 | ENST00000448348.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191100.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191100.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191100.hg.1 | ENST00000448348.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191100.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191100.hg.1 | TC0600013193.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600191096.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191096.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191096.hg.1 | ENST00000475879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191096.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191096.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191096.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191096.hg.1 | ENST00000475879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191096.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191096.hg.1 | TC0600013193.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600191101.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191101.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191101.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191101.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191101.hg.1 | TC0600013193.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600191097.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191097.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191097.hg.1 | ENST00000475879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191097.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191097.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191097.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191097.hg.1 | ENST00000475879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191097.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191097.hg.1 | TC0600013193.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367937.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000367941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000448348.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000448348.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000448348.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000448348.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000448348.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000448348.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000448348.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000475879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000475879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000475879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000475879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | ENST00000475879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | NM_003569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013193.hg.1 | NM_003569.2 | Refseq |

STX7
Genomic Map