Assay Details
| Gene Symbol: BIRC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016252.3 | NP_057336.3 | 32 - 33 | 6729 | 123 | |
| XM_005264451.4 | XP_005264508.2 | 32 - 33 | 6904 | 123 | ||
| XM_005264453.4 | XP_005264510.2 | 32 - 33 | 6607 | 123 | ||
| GenBank mRNA | AB033115.2 | - | 3 - 4 | 409 | 123 | |
| AF265555.1 | - | 32 - 33 | 6511 | 123 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000213.hg.1
Set Membership: |
|

BIRC6
Genomic Map