Assay Details
| Gene Symbol: LTBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270987.1 | NP_001257916.1 | 7 - 8 | 908 | 75 | |
| NM_002342.2 | NP_002333.1 | 7 - 8 | 1103 | 75 | ||
| GenBank mRNA | AK027080.1 | - | 7 - 8 | 968 | 75 | |
| AK095418.1 | - | 8 - 9 | 932 | 75 | ||
| AK293187.1 | - | 7 - 8 | 847 | 75 | ||
| BC026262.1 | - | 7 - 8 | 975 | 4703362 | 75 | |
| L04270.1 | - | 7 - 8 | 945 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
inflammatory response
immune response
signal transduction
multicellular organism development
viral process
response to lipopolysaccharide
tumor necrosis factor-mediated signaling pathway
regulation of cell proliferation
myeloid dendritic cell differentiation
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of JNK cascade
cellular response to mechanical stimulus
positive regulation of extrinsic apoptotic signaling pathway
tumor necrosis factor-activated receptor activity
protein binding
ubiquitin protein ligase binding
identical protein binding
apoptotic process
inflammatory response
immune response
signal transduction
multicellular organism development
viral process
response to lipopolysaccharide
tumor necrosis factor-mediated signaling pathway
regulation of cell proliferation
myeloid dendritic cell differentiation
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of JNK cascade
cellular response to mechanical stimulus
positive regulation of extrinsic apoptotic signaling pathway
tumor necrosis factor-activated receptor activity
protein binding
ubiquitin protein ligase binding
identical protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000228918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000228918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000228918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000228918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000228918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000228918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000228918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000228918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000228918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000228918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000228918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000228918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000228918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000228918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000228918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000228918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000228918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000228918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000228918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000228918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000440421.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000440421.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000440421.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000440421.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000441074.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000441074.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000441074.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000441074.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000441074.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000441074.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000441074.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000441074.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000444814.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000444814.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000444814.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000444814.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000444814.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000535739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000535739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000535739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000535739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000535739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000535739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000535739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000535739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000535739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000535739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000535739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000535739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000536876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000536876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000536876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000536876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000536876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000536876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000536876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000536876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000536876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000536876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000536876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000539925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000539925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000539925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000539925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000539925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000539925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000539925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000539925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000539925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000539925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000539925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000539925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000539925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000539925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000539925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000539925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000539925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000539925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000541005.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000541005.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000541005.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000541005.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000541005.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000541102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000541102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000541102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000541102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000541102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000541102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000541102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000541102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000541102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000541102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000541102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000542830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000542830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000542830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000542830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000542830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000542830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000542830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000542830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000543190.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000543190.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000543190.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000543190.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000543190.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000543190.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000543190.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000543190.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000543190.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000543190.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000543542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000543542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000543542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000543542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000544454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000544454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000544454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000544454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000544454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000544454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000544454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000544454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000544454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000545445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000545445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000545445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000545445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000545445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000546296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000546296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000546296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000546296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000546296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000546296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000546296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000546296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | ENST00000546296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_001270987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_001270987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_001270987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_001270987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_001270987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_001270987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_001270987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_001270987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_001270987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_001270987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_001270987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_001270987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_001270987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_001270987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_001270987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_001270987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_001270987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_001270987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_001270987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_001270987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_002342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_002342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_002342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_002342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_002342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_002342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_002342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_002342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_002342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_002342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_002342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_002342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_002342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_002342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_002342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_002342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_002342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_002342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_002342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006616.hg.1 | NM_002342.2 | Refseq |

LTBR
Genomic Map