Assay Details
| Gene Symbol: TOX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080430.3 | NP_001073899.2 | 1 - 2 | 262 | 96 | |
| XM_005255892.3 | XP_005255949.1 | 1 - 2 | 288 | 96 | ||
| GenBank mRNA | BE501808.1 | - | 1 - 2 | 262 | 96 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001115.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16015560.hg.1 | ENST00000219746.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015560.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015560.hg.1 | TC16001115.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015558.hg.1 | ENST00000219746.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015558.hg.1 | ENST00000407228.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015558.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015558.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015558.hg.1 | TC16001115.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015561.hg.1 | ENST00000219746.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015561.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015561.hg.1 | TC16001115.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600179222.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179222.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179222.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179222.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179222.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179222.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179222.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179222.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179222.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179222.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179222.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179222.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179222.hg.1 | TC1600010316.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600179226.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179226.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179226.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179226.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179226.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179226.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179226.hg.1 | TC1600010316.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600179228.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179228.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179228.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179228.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179228.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179228.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179228.hg.1 | TC1600010316.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179229.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179229.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179229.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179229.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179229.hg.1 | TC1600010316.hg.1 | Clariom D TC |

TOX3
Genomic Map