Assay Details
| Gene Symbol: TOX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080430.3 | NP_001073899.2 | 2 - 3 | 324 | 95 | |
| XM_011523002.2 | XP_011521304.1 | 2 - 3 | 1085 | 95 | ||
| XM_017023142.1 | XP_016878631.1 | 2 - 3 | 400 | 95 | ||
| GenBank mRNA | AK027006.1 | - | 1 - 2 | 59 | 95 | |
| BE501808.1 | - | 2 - 3 | 324 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001115.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015556.hg.1 | ENST00000219746.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015556.hg.1 | ENST00000407228.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015556.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015556.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015556.hg.1 | TC16001115.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600179220.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179220.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179220.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179220.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179220.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179220.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179220.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179220.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179220.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179220.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179220.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179220.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179220.hg.1 | TC1600010316.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 125060 | - | - |

TOX3
Genomic Map