Assay Details
| Gene Symbol: TOX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080430.3 | NP_001073899.2 | 3 - 4 | 575 | 107 | |
| NM_001146188.2 | NP_001139660.1 | 4 - 5 | 577 | 107 | ||
| XM_005255892.3 | XP_005255949.1 | 3 - 4 | 598 | 107 | ||
| XM_011523002.2 | XP_011521304.1 | 3 - 4 | 1336 | 107 | ||
| XM_017023142.1 | XP_016878631.1 | 3 - 4 | 651 | 107 | ||
| GenBank mRNA | AK299202.1 | - | 4 - 5 | 577 | 107 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001115.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16015555.hg.1 | ENST00000219746.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015555.hg.1 | ENST00000407228.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015555.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015555.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015555.hg.1 | TC16001115.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015559.hg.1 | ENST00000407228.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015559.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015559.hg.1 | TC16001115.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015562.hg.1 | ENST00000407228.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015562.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015562.hg.1 | TC16001115.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600179218.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179218.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179218.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179218.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179218.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179218.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179218.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179218.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179218.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179218.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179218.hg.1 | TC1600010316.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600179225.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179225.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179225.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179225.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179225.hg.1 | TC1600010316.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600179232.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179232.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179232.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179232.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179232.hg.1 | TC1600010316.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000566696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | ENST00000568436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010316.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179224.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179224.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179224.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179224.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179224.hg.1 | TC1600010316.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600179219.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179219.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179219.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179219.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179219.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179219.hg.1 | ENST00000219746.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179219.hg.1 | ENST00000407228.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179219.hg.1 | ENST00000563091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179219.hg.1 | NM_001080430.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179219.hg.1 | NM_001146188.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179219.hg.1 | TC1600010316.hg.1 | Clariom D TC |

TOX3
Genomic Map