Assay Details
| Gene Symbol: DPCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015448.1 | NP_056263.1 | 2 - 3 | 155 | 83 | |
| GenBank mRNA | AK290154.1 | - | 2 - 3 | 156 | 83 | |
| AL110240.1 | - | 1 - 2 | 76 | 83 | ||
| AY532267.1 | - | 2 - 3 | 142 | 83 | ||
| BC001082.2 | - | 2 - 3 | 155 | 2907119 | 83 | |
| BC031695.1 | - | 2 - 3 | 178 | 5166605 | 83 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000739.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000370147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000370147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000370147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000370147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000370147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000370147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000370147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000370147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000370147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000370151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000370151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000370151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000370151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000370151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000370151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000370151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000370151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000370151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000370151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000434727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000434727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000434727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000434727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000470165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000470165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000470165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000470165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000470165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000470165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000475443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000475443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000475443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000475443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000475443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000475443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000475443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000626968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000626968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000626968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000626968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000626968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000626968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | ENST00000626968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | NM_015448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | NM_015448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | NM_015448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | NM_015448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | NM_015448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | NM_015448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | NM_015448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | NM_015448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | NM_015448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008686.hg.1 | NM_015448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008744.hg.1 | ENST00000370147.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008744.hg.1 | ENST00000370148.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008744.hg.1 | ENST00000370149.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008744.hg.1 | ENST00000370151.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008744.hg.1 | NM_015448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008744.hg.1 | TC10000739.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000159398.hg.1 | ENST00000370147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159398.hg.1 | ENST00000370151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159398.hg.1 | ENST00000434727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159398.hg.1 | ENST00000475443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159398.hg.1 | ENST00000626968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159398.hg.1 | NM_015448.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000159398.hg.1 | ENST00000370147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159398.hg.1 | ENST00000370151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159398.hg.1 | ENST00000434727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159398.hg.1 | ENST00000475443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159398.hg.1 | ENST00000626968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159398.hg.1 | NM_015448.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000159398.hg.1 | TC1000008686.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000370147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000370147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000370147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000370147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000370147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000370147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000370147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000370147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000370147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000370151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000370151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000370151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000370151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000370151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000370151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000370151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000370151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000370151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000370151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000434727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000434727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000434727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000434727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000470165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000470165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000470165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000470165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000470165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000470165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000475443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000475443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000475443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000475443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000475443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000475443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000475443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000626968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000626968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000626968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000626968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000626968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000626968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | ENST00000626968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | NM_015448.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | NM_015448.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | NM_015448.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | NM_015448.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | NM_015448.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | NM_015448.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | NM_015448.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | NM_015448.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | NM_015448.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008686.hg.1 | NM_015448.2 | Refseq |

DPCD
Genomic Map