Assay Details
| Gene Symbol: HLA-DOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK301404.1 | - | 1 - 2 | 128 | 107 | |
| M38056.1 | - | 1 - 2 | 134 | 107 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK301404.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000374813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000374813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000374813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000374813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000467465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000467465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000467465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000467465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000467465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000485901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000485901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000485901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000485901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000485901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000485901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000485901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000485901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000485901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000490305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000490305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000490305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000490305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000490305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000495532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000495532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000495532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000495532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000495532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | ENST00000495532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000229829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000374813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000374813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000374813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000374813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000450833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000467465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000467465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000467465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000467465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000467465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000485901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000485901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000485901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000485901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000485901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000485901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000485901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000485901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000485901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000490305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000490305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000490305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000490305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000490305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000495532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000495532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000495532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000495532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000495532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | ENST00000495532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | NM_002119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011516.hg.1 | NM_002119.3 | Refseq |

HLA-DOA
Genomic Map