| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029602.hg.1 |
TC02001855.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029602.hg.1 |
ENST00000317610.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029602.hg.1 |
ENST00000337526.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029602.hg.1 |
ENST00000354474.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029602.hg.1 |
ENST00000357376.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029602.hg.1 |
ENST00000357732.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029602.hg.1 |
ENST00000394609.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029602.hg.1 |
ENST00000394611.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029602.hg.1 |
ENST00000402434.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029602.hg.1 |
ENST00000404909.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029602.hg.1 |
ENST00000405240.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029602.hg.1 |
NM_007008.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029602.hg.1 |
NM_020532.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029602.hg.1 |
NM_153828.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029602.hg.1 |
NM_207520.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029602.hg.1 |
NM_207521.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
ENST00000317610.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
ENST00000337526.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
ENST00000357376.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
ENST00000357732.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
ENST00000394609.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
ENST00000394611.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
ENST00000402434.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
ENST00000404909.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
ENST00000405240.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
ENST00000438462.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
NM_007008.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
NM_020532.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
NM_153828.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
NM_207520.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
NM_207521.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
ENST00000317610.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
ENST00000337526.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
ENST00000357376.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
ENST00000357732.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
ENST00000394609.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
ENST00000394611.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
ENST00000402434.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
ENST00000404909.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
ENST00000405240.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
ENST00000438462.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
NM_007008.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
NM_020532.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
NM_153828.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
NM_207520.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
NM_207521.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200226416.hg.1 |
TC0200016672.hg.1 |
Clariom D TC |